Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Procalcitonin Monoclonal Antibody (42)
GREENER_CHOICE

Catalog No. MA120289
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA120289 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA120289 Supplier Invitrogen™ Supplier No. MA120289
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-20289 detects procalcitonin in human samples.

This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Procalcitonin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 42
Concentration 6.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CALCA
Gene Accession No. P01258, P06881
Gene Alias alpha-type CGRP; CALC1; CALCA; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCP; CGRP; CGRP1; CGRP-I; CT; Katacalcin; KC; PCT; PDN-21
Gene Symbols CALCA
Host Species Mouse
Immunogen Fragments of human PCT conjugated with carrier protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.