Learn More
Description
ProClean™ garments are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other cleanroom or controlled critical environments.
Specifications
Specifications
| Size | Large |
| Gender | Unisex |
| Material | SMS |
| Color | Blue |
| No. of Pockets | 1 |
| Sterility | Non-sterile |
| For Use With (Application) | Controlled Environments |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.