Learn More
DuPont™ ProClean™ Limited-Use Cleanroom Coverall with Hood and Boots
Shop All DuPont Personal Protection ProductsDescription
- Full-length zippered front
- Attached hood and boots with elastic face opening
- Tunnelized, snug-fitting elastic at the face, wrists, ankles, and back-waist
Order Info
Available with no special cleaning (bulk) or cleaned, in a variety of sizes.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Large |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Description | Finish: Clean |
| For Use With (Application) | Controlled Environments |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.