Learn More
Alkali Scientific ProCycle™ 0.2mL 12-strip, PCR Tubes with Dome Caps, Assorted Colors, 80 Strips/Pack
SDP
Supplier:  Alkali Scientific PC7058
Encompass_Preferred
Assorted Colors Angularly attached caps on UltraFlux i and SnapStrip tubes prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack. Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing. Convenient combo pack of tubes and caps -- no need to purchase separately Colored tubes offer easy organization. Dual-colored with optically-clear flat caps to ensure that the fluorescence signal is uninhibited for optimum qPCR performance. Assorted colors are individually packed  For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-302-8294
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.