Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Prodan, Membrane Surface Probe, 227.3 Da, 25 mg

Catalog No. 502705363
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5363 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5363 Supplier Abcam Supplier No. AB14524625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent membrane surface probe for studying phospholipid interactions in in vitro assays.

  • Molecular Weight: 227.3 Da
  • Purity: >98%
  • Application: Membrane surface probe; suitable for use in non-cell based in vitro assays or membrane models for studying phospholipid interactions
  • Storage Buffer: 100% Prodan

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Phospholipids
Substrate Properties Chemical Substrate
Shipping Condition Blue Ice
Buffer 100% Prodan
Content And Storage Store at -20°C.
Formulation 100% Prodan
Quantity 25 mg
For Use With (Application) Non-cell based in vitro assays, membrane models for studying phospholipid interactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.