Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human BMP-4 Recombinant Protein

Catalog No. PRD31410
Change view
Click to view available options
Quantity:
250 μg
5 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD31410 50 μg
PRD31401 5 μg
PRD31450 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PRD31410 Supplier R&D Systems™ Supplier No. PRD31410
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human BMP-4 Protein is derived from NS0. The ProDots Recombinant Human BMP-4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q53XC5
For Use With (Application) Bioactivity
Gene ID (Entrez) 652
Molecular Weight (g/mol) Observed M.W.: 22-25kDa, reducing conditions 37-41, non-reducing conditions, Theoretical M.W.: 13kDa (monomer)
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human BMP-4 protein Ser293-Arg408
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method.
Gene Alias BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Common Name BMP-4
Gene Symbol BMP4
Biological Activity Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is typically 2.5-15.0ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 50-200 μg/mL in sterile 4 mM HCl, or simply roll ProDot™ directly into cell culture medium for immediate use.
Expression System Mouse myeloma cell line, NS0-derived Ser293 - Arg408
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.