Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human IL-3 Recombinant Protein

Catalog No. PRD20350
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD20350 250 μg
PRD20310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD20350 Supplier R&D Systems™ Supplier No. PRD20350
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human IL-3 Protein is derived from E. coli. The ProDots Recombinant Human IL-3 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q6GS87
For Use With (Application) Bioactivity
Gene ID (Entrez) 3562
Molecular Weight (g/mol) Predicted MW: 15kDa Observed MW: 14kDa, reducing conditions
Quantity 250 μg
Source E. coli-derived human IL-3 protein Ala20-Phe152,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor
Common Name IL-3
Gene Symbol IL3
Biological Activity Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is typically 0.02-0.1ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS, or simply roll ProDot directly into cell culture medium for immediate use.
Expression System E. coli-derived Ala20 - Phe152, with and without an N-terminal Met
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.