Learn More
R&D Systems™ ProDots Recombinant Human IL-6 Protein
Description
The ED50 for this effect is 0.2-0.8 ng/mL.
Specifications
Specifications
| Accession Number | Q75MH2 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | M.W. (Observed): 21 kDa, reducing conditions; M.W. (theoretical): 20.3 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived human IL-6 protein Pro29 - Met212 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2°C to 8°C as supplied. 1 month at 2°C to 8°C under sterile conditions after reconstitution. 3 months (at -20°C to -80°C under sterile conditions after reconstitution.) |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A |
| Common Name | IL-6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.