Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human IL-6 Protein

Catalog No. PRD20650
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD20650 250 μg
PRD20610 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD20650 Supplier R&D Systems™ Supplier No. PRD20650
This item is not returnable. View return policy

Supplied as ready-to-use 5 ug protein balls

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813.

The ED50 for this effect is 0.2-0.8 ng/mL.

Specifications

Accession Number Q75MH2
For Use With (Application) Bioactivity
Gene ID (Entrez) 3569
Molecular Weight (g/mol) M.W. (Observed): 21 kDa, reducing conditions; M.W. (theoretical): 20.3 kDa
Quantity 250 μg
Source E. coli-derived human IL-6 protein Pro29 - Met212
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2°C to 8°C as supplied. 1 month at 2°C to 8°C under sterile conditions after reconstitution. 3 months (at -20°C to -80°C under sterile conditions after reconstitution.)
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Common Name IL-6
Gene Symbol IL6
Biological Activity Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is typically 0.2-0.8ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS, or simply roll ProDot directly into cell culture medium for immediate use.
Expression System E. coli-derived Pro29 - Met212
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.