Learn More
R&D Systems™ ProDots Human PDGF-BB Recombinant Protein
Description
Specifications
Specifications
| Accession Number | Q6FHE7 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 5155 |
| Molecular Weight (g/mol) | Observed M.W.: 12kDa, reducing conditions, Theoretical M.W.: 12.3kDa (monomer) |
| Quantity | 5 μg |
| Source | E. coli-derived human PDGF-BB protein Ser82-Thr190 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Common Name | PDGF-BB |
| Gene Symbol | PDGFB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.