Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots™ Human Recombinant FGF-3 Protein

Catalog No. PRD120610
Change view
Click to view available options
Quantity:
15 μg
5 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD120610 50 μg
PRD120601 5 μg
PRD120650 15 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PRD120610 Supplier R&D Systems™ Supplier No. PRD120610
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human FGF-3 Protein is derived from E. coli. The ProDots Recombinant Human FGF-3 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

The product is shipped at ambient temperature.

Specifications

Accession Number Q13241
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 2248
Molecular Weight (g/mol) 21.1 kDa, reducing conditions
Name FGF3, fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, Hst, INT-2, mouse, oncogene INT2
Quantity 50 μg
Source E. coli-derived human FGF-3 protein Asp28-Arg212,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity 0.2 to 1μg/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Protein Tag No
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS, or simply roll ProDot™ directly to the medium for immediate use.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.