Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Human VEGF 165 Recombinant Protein

Catalog No. PRD29301
Change view
Click to view available options
Quantity:
5 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD29301 5 μg
PRD29310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD29301 Supplier R&D Systems™ Supplier No. PRD29301
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human VEGF 165 Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human VEGF 165 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_001165097
For Use With (Application) Bioactivity
Gene ID (Entrez) 7422
Molecular Weight (g/mol) Observed M.W.: 20-22kDa, reducing conditions 39-42kDa, non-reducing conditions, Theoretical M.W.: 19.2kDa (monomer)
Quantity 5 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human VEGF protein Ala27-Arg191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Common Name VEGF
Gene Symbol VEGFA
Biological Activity Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 is typically 1.0-6.0ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS or (at 100-500 μg/mL in sterile 4 mM HCl. For immediate use, simply roll ProDot™ directly into cell culture medium.)
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Ala27 - Arg191
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.