Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human Flt-3 Ligand Protein

Catalog No. PRD30801
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD30801 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PRD30801 Supplier R&D Systems™ Supplier No. PRD30801
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The ProDots Recombinant Human Flt-3 Ligand Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human Flt-3 Ligand Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAA17999.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.5 kDa
Quantity 5 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Flt-3 Ligand protein Thr27-Pro185
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8 °C as supplied. 1 month at 2-8 °C under sterile conditions after reconstitution. 3 months at -20 to -80 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Gene Symbol FLT3LG
Specific Reactivity Flt-3 Ligand
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS, or simply roll ProDot« directly into cell culture medium for immediate use.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.