Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots™ Recombinant Human IL-1 beta/IL-1F2 Protein

Catalog No. PRD20110
Change view
Click to view available options
Quantity:
5 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD20110 50 μg
PRD20101 5 μg
PRD20150 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PRD20110 Supplier R&D Systems™ Supplier No. PRD20110
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human IL-1 beta/IL-1F2 Protein is derived from E. coli. The ProDots Recombinant Human IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number NP_000567
For Use With (Application) Bioactivity
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17 kDa
Name catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Quantity 50 μg
Source E. coli-derived human IL-1 beta/IL-1F2 protein Ala117-Ser269
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Biological Activity Measured in a cell proliferation assay using D10.G4.1 mouse helper T cells. Symons, J.A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. (eds): IRL Press. 272. The ED50 for this effect is typically <12 pg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 25 μg/mL in sterile PBS, or simply roll ProDot™ directly into cell culture medium for immediate use.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.