Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human R-Spondin 1 Protein

Catalog No. PRD464510
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD464510 50 μg
PRD464550 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD464510 Supplier R&D Systems™ Supplier No. PRD464510
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The ProDots Recombinant Human R-Spondin 1 Protein is derived from CHO. The ProDots Recombinant Human R-Spondin 1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q2MKA7
Characterization Bioactivity
For Use With (Application) Bioactivity
Gene ID (Entrez) 284654
Molecular Weight (g/mol) 25.6kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human R-Spondin 1 protein Arg31-Ala263
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution For a stock solution, reconstitute at 100 μg/mL in sterile PBS. For immediate use, simply roll ProDot™ directly into cell culture medium.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.