Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container, BioTitan retainers, three ports, MPX and Luer locks

Description
Productainer BPCs are constructed with our Thermo Scientific Aegis5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.
Features include:
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
- Now manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
Recommended for:
- Harvest—collection and storage of bulk product from bioreactors
- Storage—sterile storage of media, buffers and process intermediates
- Separation—feeding and receiving of sterile liquids from separation systems
- Purification—buffer feeding to purification system, fraction collection and storage
- Filling—sterile reservoir for filling systems
- Transport and Shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Capacity (Metric) | 100 L |
| For Use With (Application) | Single-Use Bioprocessing |
| Quantity | 100 L |
| Tubing Style | TPE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.