Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Prodynorphin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA127974 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA127974 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA127974 Supplier Invitrogen™ Supplier No. PA127974
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Guinea Pig Polyclonal Antibody

Expected to react with mouse samples due to 100% sequence homology.

expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prodynorphin
Applications Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.05% sodium azide
Gene Pdyn
Gene Accession No. O35417, P06300
Gene Alias ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product
Gene Symbols Pdyn
Host Species Guinea Pig
Immunogen SQENPNTYSEDLDV. Corresponding to residues 235-248 of the rat proDynorphin.
Purification Method Affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18610, 29190
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.