Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Professional HEALTHY SOAP™
SDP

Catalog No. 19043833 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
PURELL ES4 Push-Style Soap Dispensers
PURELL ES6 Touch-Free Soap Dispensers
PURELL ES8 Touch-Free Soap Dispensers
PURELL FMX-20 Push-Style Hand Sanitizer Dispensers
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-043-833 PURELL ES6 Touch-Free Soap Dispensers
19-043-823 PURELL FMX-20 Push-Style Hand Sanitizer Dispensers
19-043-834 PURELL ES8 Touch-Free Soap Dispensers
19-043-851 PURELL ES4 Push-Style Soap Dispensers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-043-833 Supplier Gojo Industries Inc Supplier No. 647902

Professional HEALTHY SOAP™ is a quick-acting antimicrobial foam soap for general handwashing.

  • Efficacy and mildness come together in this effective antimicrobial hand soap
  • Quick-acting antimicrobial foam soap for general handwashing
  • Proven to be as mild as our top selling non-antimicrobial soap
  • Hypoallergenic
  • Dermatologist tested
  • Maintains the skin natural moisture barrier
  • Dye-Free with light fragrance
  • AT-A-GLANCE™ refills monitor product level with just one look
  • SANITARY SEALED™ PET refill is easily recycled
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Soap
Color Blue
Form Foam
For Use With (Application) Hand Washing
For Use With (Equipment) PURELL ES6 Touch-Free Soap Dispensers
Quantity 2/Pk.
Size 1200 mL
Volume (English) 40.58 oz.
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.