Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 100HV Economy Spandex Hi-Vis Back Support Brace

Catalog No. 19819470 Shop All Ergodyne Products
Change view
Click to view available options
Size:
X-Small
Small
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-470 2X-Large
19-819-468 X-Small
19-819-469 Small
19-819-471 3X-Large
19-819-472 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-819-470 Supplier Ergodyne™ Supplier No. 11886
Add to Cart
Add to Cart

ProFlex™ 100HV Economy Spandex Hi-Vis Back Support designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.

  • High visibility
  • Polypropylene stays for long-lasting integrity
  • Adjustable and removable 0.5 to 1 in. (1 to 2.5 cm) shoulder straps keep the belt on standby until the next lift
  • Stretchable bias binding with zig-zag stitching
  • Rubber track webbing prevents riding up
  • Machine wash cold, air dry

Specifications

Color Orange
Size 2X-Large
For Use With (Application) Material Handling, Construction, Maintenance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.