Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 1505 Low-Profile Weight Lifters Back Support Brace

Catalog No. 19819462 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-462 Small
19-819-464 Medium
19-819-465 Large
19-819-466 X-Large
19-819-467 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-819-462 Supplier Ergodyne™ Supplier No. 11491

Low-Profile Weight Lifters Back Support with flexible soft foam core provides support when lifting.

  • 6.25 in. low-profile tapered design
  • Flexible soft foam core composition
  • Nylon jersey body fabric
  • Easy cam buckle closure system
  • Rubber track webbing

Specifications

Color Black
Size Small
For Use With (Application) Material Handling, Construction, Warehousing/Distribution, Maintenance
Width (English) 6.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.