Learn More
Description
- 6.25 in. low-profile tapered design
- Flexible soft foam core composition
- Nylon jersey body fabric
- Easy cam buckle closure system
- Rubber track webbing
Specifications
Specifications
| Color | Black |
| Size | 2X-Large |
| For Use With (Application) | Material Handling, Construction, Warehousing/Distribution, Maintenance |
| Width (English) | 6.25 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.