Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 1600 Standard Elastic Back Support Brace

Catalog No. 19819452 Shop All Ergodyne Products
Change view
Click to view available options
Size:
X-Small
Large
3X-Large
4X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-819-452 Large
19-819-451 X-Small
19-819-454 4X-Large
19-819-453 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-819-452 Supplier Ergodyne™ Supplier No. 11104

ProFlex™ 1600 Standard Elastic Back Support is designed to remind workers of proper lifting techniques while moving stress away from the spine for back pain relief.

  • Premium 9 in. (22.9 cm) all-elastic body provides firm support
  • Contoured high-cut front panel for a comfortable fit
  • Polypropylene for long-lasting integrity
  • Adjustable, detachable suspenders keep belt in place
  • Rubber track webbing prevents riding up
  • Waist size ranges from 25 to 58 in. (64 to 147 cm)
  • Machine wash cold, air dry

Specifications

Color Black
Size Large
For Use With (Application) Material Handling, Construction,Warehousing/Distribution, Maintenance
Includes Suspenders
Width (English) 9 in.
Width (Metric) 22.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.