Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 365 Mini Kneeling Pad

ProFlex™ 365 Mini Kneeling Pad constructed of nitrile rubber (NBR) closed cell foam is designed to keep its shape over time.

$39.08

Specifications

Color Black
Dimensions (L x W) 4 x 6 in. (10 x 15 cm)
For Use With (Application) Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Material Nitrile Butadiene Rubber
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-820-281
View Documents
Ergodyne™
18465
Grabber
Each for $39.08
Only null left
Add to Cart
 
19-820-290
View Documents
Ergodyne™
18565
Carabiner
Each for $39.08
Only null left
Add to Cart
 
Description

Description

  • Small and lightweight for optimal portability and storage
  • Built-in handle and carabiner for hanging storage or easy attachment
  • 1 in. (25 mm) NBR foam padding is nonconductive and keeps its shape over time
  • Water resistant and petroleum resistant construction
  • Use to support and alleviate pressure from other areas like elbows, shoulders, and hips
Specifications

Specifications

Black
4 x 6 in. (10 x 15 cm)
Flagging Crew, Road Construction, Utility Crew, Survey Crew, Construction
Nitrile Butadiene Rubber
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.