Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 900 Half-Finger Impact Gloves

Catalog No. 19820201 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-201 X-Large
19-820-198 Small
19-820-199 Medium
19-820-200 Large
19-820-202 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-201 Supplier Ergodyne™ Supplier No. 17695
Add to Cart
Add to Cart
This item is not returnable. View return policy

AVC™ padding in the palm and other key areas helps to reduce shock and impact. Coating on the palm provides enhanced gripping capabilities.

  • Segmented AVC™ palm padding reduces shock and palm impact
  • Gripping palm provides secure hold on tools and equipment
  • Half-finger design for dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with a reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves include ID space to write user's names
  • Finger webbing loops for easy on/off

Specifications

Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size X-Large
For Use With (Application) Construction, Manufacturing, Hand and Power Tools
Glove Cuff Style Hook and Loop
Series ProFlex™ 900
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.