Learn More
Description
- Segmented AVC™ padding reduces shock and impact
- Gripping palm provides a secure hold on tools and equipment
- Full-finger design enhances protection while maintaining dexterity
- Hex stretch knit and breathable spandex construction
- Neoprene knuckle pad with a reinforced thumb saddle
- Low-profile molded hook & loop closure
- Gloves feature ID space to write user's name
Specifications
Specifications
| Cuff Material | Neoprene |
| Color | Black |
| Hand | Specific |
| Material | Spandex |
| Product Type | Anti-impact Gloves |
| Size | Medium |
| For Use With (Application) | Oil/Gas Drilling, Mining, and Heavy Construction |
| Glove Cuff Style | Hook and Loop |
| Series | ProFlex™ 9001 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.