Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 9001 Full-Finger Impact Gloves with AVC™ Padding

Catalog No. 19820232 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-232 X-Large
19-820-219 Small
19-820-229 Medium
19-820-230 Large
19-820-233 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-232 Supplier Ergodyne™ Supplier No. 17775
Add to Cart
Add to Cart
This item is not returnable. View return policy

AVC™ padding in the palm and other key areas helps to reduce the effect of shocks and impacts. Full finger design enhances protection without sacrificing dexterity.

  • Segmented AVC™ padding reduces shock and impact
  • Gripping palm provides a secure hold on tools and equipment
  • Full-finger design enhances protection while maintaining dexterity
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with a reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Cuff Material Neoprene
Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size X-Large
For Use With (Application) Oil/Gas Drilling, Mining, and Heavy Construction
Glove Cuff Style Hook and Loop
Series ProFlex™ 9001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.