Learn More
Description
- Segmented foam palm padding reduces shock and impact
- Genuine leather palm provides durability with a natural feel
- Half-finger design for dexterity and tactile feedback
- Hex stretch knit and breathable spandex construction
- Finger webbing loops for easy on/off
- Neoprene knuckle pad
- Low-profile molded hook & loop closure
- Gloves feature ID space to write user's name
Specifications
Specifications
| Back Material | Spandex |
| Color | Black |
| Hand | Specific |
| Material | Geniune Leather |
| Product Type | Anti-impact Gloves |
| Size | X-Large |
| For Use With (Application) | Construction, Manufacturing, Hand and Power Tools |
| Glove Cuff Style | Hook and Loop |
| Series | ProFlex™ 901 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.