Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 9015F(x) ANSI/ISO-Certified Anti-Vibration Gloves with DIR Protection

Catalog No. 19820147 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-147 Medium
19-820-146 Small
19-820-148 Large
19-820-149 X-Large
19-820-150 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-147 Supplier Ergodyne™ Supplier No. 17303
Add to Cart
Add to Cart
This item is not returnable. View return policy

Full-length AVC™ palm padding reduces vibration and palm impact, while molded TPR armor on the knuckles protects the back of the hand.

  • ANSI S2.73-2014/ISO 10819:2013-certified anti-vibration protection
  • Full-length AVC™ palm padding reduces vibration and palm impact
  • Molded TPR armor on the knuckles and fingers protects the back of the hand
  • Palm coating enhances grip for a secure hold on tools and equipment
  • Hex stretch knit, breathable spandex construction
  • Low-profile molded hook & loop closure
  • ID space for user personalization
  • Reinforced thumb saddle

Specifications

Back Material TPR Padding
Certifications/Compliance ANSI S2.73-2014/ISO 10819:2013
Color Black
Hand Specific
Material Spandex
Product Type Anti-vibration Gloves
Size Medium
EN388 Standard EN388:2121X
For Use With (Application) Assembly, Demolition, Heavy Construction, and Oil/Gas Refining
Glove Cuff Style Hook and Loop
Palm Type AVC™ Padding
Series ProFlex™ 9015F(x)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.