Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 910 Half-Finger Impact Gloves with Wrist Support

Catalog No. 19820212 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-212 X-Large
19-820-209 Small
19-820-210 Medium
19-820-211 Large
19-820-213 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-212 Supplier Ergodyne™ Supplier No. 17715
Add to Cart
Add to Cart
This item is not returnable. View return policy

AVC™ padding in the palm and other key areas helps to reduce shock and impact. Built-in wrist support with Open-Center Stay™ technology relieves discomfort caused by repetitive motion.

  • Segmented AVC™ palm padding reduces shock and palm impact
  • Coated palm provides enhanced grip capabilites
  • Built-in wrist support with patented Open-Center Stay™ aides in relief from repetitive motion discomfort
  • Half-finger design for maximum dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Cuff Material Neoprene
Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size X-Large
For Use With (Application) Construction, Manufacturing, Repetative Motion Tasks, Hand and Power Tools
Glove Cuff Color Black
Glove Cuff Style Hook and Loop
Series ProFlex™ 910
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.