Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 924 Hybrid Dorsal Impact-Reducing Gloves

Catalog No. 19820194 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-194 Medium
19-820-193 Small
19-820-195 Large
19-820-196 X-Large
19-820-197 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-194 Supplier Ergodyne™ Supplier No. 17683
This item is not returnable. View return policy

Gloves feature thermoplastic rubber construction throughout the fingers, with flexible foam knuckle padding for additional impact protection.

  • Padded palm reinforced with Tena-Grip™ technology
  • Hi-vis breathable mesh construction
  • TPR fingertips with foam knuckle padding and a reinforced thumb saddle
  • Neoprene cuff with molded hook and loop closure keeps out debris
  • Gloves feature ID space to write user's name

Specifications

Back Material Mesh
Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Medium
For Use With (Application) Oil/Gas Drilling, Mining, Construction, and Mechanics
Glove Cuff Style Hook and Loop
Series ProFlex™ 924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.