Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 924LTR Leather-Reinforced Hybrid Dorsal Impact-Reducing Gloves

Catalog No. 19820242 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-242 Large
19-820-240 Small
19-820-241 Medium
19-820-243 X-Large
19-820-245 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-242 Supplier Ergodyne™ Supplier No. 17794
This item is not returnable. View return policy

Gloves feature thermoplastic rubber construction throughout the fingers with a fully padded, genuine leather reinforced palm.

  • Leather-reinforced fingers and thumb saddle
  • Hi-vis breathable mesh construction
  • Neoprene cuff with molded hook and loop closure keeps out debris
  • Gloves feature ID space to write user's names

Specifications

Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Large
For Use With (Application) Roofing, Mining, Oil/Gas Drilling, Construction, and Mechanics
Glove Cuff Style Hook and Loop
Series ProFlex™ 924LTR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.