Learn More
Ergodyne™ ProFlex™ 924LTR Leather-Reinforced Hybrid Dorsal Impact-Reducing Gloves
Shop All Ergodyne ProductsDescription
- Leather-reinforced fingers and thumb saddle
- Hi-vis breathable mesh construction
- Neoprene cuff with molded hook and loop closure keeps out debris
- Gloves feature ID space to write user's names
Specifications
Specifications
| Cuff Material | Neoprene |
| Color | Hi-vis Lime Green, Black |
| Hand | Specific |
| Material | Thermoplastic Rubber |
| Product Type | Impact-Reducing Gloves |
| Size | 2X-Large |
| For Use With (Application) | Roofing, Mining, Oil/Gas Drilling, Construction, and Mechanics |
| Glove Cuff Style | Hook and Loop |
| Series | ProFlex™ 924LTR |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.