Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 925CR6 Performance Dorsal Impact-Reducing and Cut Resistant Gloves

Catalog No. 19820141 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-141 Small
19-820-142 Medium
19-820-143 Large
19-820-144 X-Large
19-820-145 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-141 Supplier Ergodyne™ Supplier No. 17292
This item is not returnable. View return policy

Impact-reducing and cut-resistant gloves with hi-visibility suede and non-slip dot grip palm provide maximum impact protection and full coverage.

  • Next-gen Armortex™-backed palm for extreme cut protection
  • Full TPR armor and technical foam for max protection
  • Hi-Vis AX Suede™ and non-slip dot gripping palm
  • Armortex™-reinforced thumb saddle and index finger
  • Padded neoprene cuff with reflective binding
  • Pull-on tab with ID space

Specifications

Color Lime Green
Cut Resistance Rating ANSI A6
Material Next-gen Armortex
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4443
For Use With (Application) Oil/Gas Drilling, Mining, Heavy Construction, Ironwork
Glove Cuff Color Black, Gray
Glove Cuff Style Padded neoprene cuff with reflective binding
Liner Material TPR Armor, Technical Foam
Model 17292
Series ProFlex™ 925CR6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.