Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 925F(x) Standard Dorsal Impact-Reducing Gloves

Catalog No. 19820254 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-254 Medium
19-820-253 Small
19-820-255 Large
19-820-256 X-Large
19-820-257 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-254 Supplier Ergodyne™ Supplier No. 17903
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are constructed of full thermoplastic rubber armor with technical foam padding for additional protection. Palms feature Hi-Vis AX Suede™ non-slip dots for enhanced gripping capabilities.

  • Full TPR armor with technical foam padding
  • Hi-Vis AX Suede™ with non-slip dot gripping palm
  • Armortex™-reinforced thumb saddle and index finger
  • Hi-vis breathable mesh construction
  • Padded neoprene cuff with reflective binding
  • Pull-on tab with ID space to write user's name

Specifications

Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Medium
For Use With (Application) Oil/Gas Drilling, Mining, and Heavy Construction
Glove Cuff Style Open
Series ProFlex™ 925F(x)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.