Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 4010 Double Strap Wrist Support

Catalog No. 19910439 Shop All Ergodyne Products
Change view
Click to view available options
Hand:
Left
Right
Color:
Black
Tan
Size:
Small
Medium
Large
X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand Color Size
19-910-439 Right Tan Medium
19-910-432 Right Black Small
19-910-442 Left Tan Small
19-910-443 Left Tan Medium
19-910-444 Left Tan Large
19-910-440 Right Tan Large
19-910-445 Left Tan X-Large
19-910-441 Right Tan X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-910-439 Supplier Ergodyne™ Supplier No. 70124

Double strap wrist support with open center supports the wrist during repetitive motion discomfort by limiting wrist movements.

Specifications

Color Tan
Size Medium
For Use With (Application) Food Processing, Picking/Packing,Assembly/Fabrication
Hand Right
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.