Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 819WP Extreme Thermal Waterproof Winter Work Gloves

Catalog No. 19820184 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-184 Large
19-820-182 Small
19-820-183 Medium
19-820-185 X-Large
19-820-186 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-184 Supplier Ergodyne™ Supplier No. 17404
This item is not returnable. View return policy

Gloves combine dual-zone 3M™ Thinsulate™ Insulation with a waterproof, windproof breathable membrane insert for long lasting comfort and protection.

  • Dual-zone 3M™ Thinsulate™ Insulation for cold weather protection
  • Waterproof, windproof breathable membrane insert
  • Weather-resistant ripstop outer shell with DWR finish
  • Rugged AX Suede™ palm and fingertip reinforcements
  • Neoprene knuckle pad protects the back of the hand
  • Reflective accents increase worker safety in low light applications
  • Touchscreen capable thumb and index finger
  • Gauntlet cuff with internal elastic cinch cord
  • Soft thumb nose/brow wipe area

Specifications

Back Material Neoprene
Color Black
Hand Specific
Size Large
EN388 Standard 3423 X
For Use With (Application) Cold Weather Work, Cold Storage/Refrigeration, Construction, Mining, and Outdoor Recreation
Glove Cuff Style Gauntlet
Liner Material 3M™ Thinsulate™
Palm Type AX Suede™
Series ProFlex™ 819WP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.