Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 7041 Hi-Vis Nitrile-Coated Cut-Resistant Gloves

Catalog No. 19820250 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-250 Large
19-820-248 Small
19-820-249 Medium
19-820-251 X-Large
19-820-252 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-250 Supplier Ergodyne™ Supplier No. 17814
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed with two new technologies to create the a comfortable, breathable and gripping ANSI cut level 4 gloves. Ideal for workers in construction, assembly, fabrication, pulp and paper, service technicians or workers handling sheet metal or other sharp edge materials.

  • Made with high-performance polyethylene (HPPE) fibers for ANSI 4 cut protection
  • HPPE does not include steel or glass often used in other anti-cut gloves which can cause irritation
  • WSX™ dip, combined with TenaLux™ yarn make these palm-coated gloves cool to the touch with 360-degrees of breathability
  • Lighter weight than traditional A4 cut-resistant gloves
  • Hi-vis lime color on the backside of the hand
  • Liquid-resistant special coating absorbs and wicks moisture from the surface allowing for a firm-grip in wet oily and wet conditions
  • Water-based polyurethane (PU) coating combined with nitrile dip allows maximum dexterity and significantly relieves hand fatigue while ensuring durability

Specifications

ANSI Abrasion Rating ANSI 4
Construction Knit
Color Hi-vis Lime Green
Cut Resistance Rating ANSI A4
Material HPPE
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Construction, assembly, fabrication, pulp and paper, service technicians
Glove Cuff Style Knit
Grip Design WSX dip technology
Model 17814
Series ProFlex™ 7041
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.