Description
- Ultra-durable hand protection for handling sharp or jagged materials
- Tena-Grip™ reinforcements on the padded palm, fingertips, and thumb saddle
- Technical-hex stretch knit material with a neoprene knuckle pad to protect the back of the hand
- Hook & Loop closure provides a secure, customizable fit
- Enhanced grip in wet or dry applications
- Pull-on tab enables easy on and off
- ID space allows for glove personalization
Specifications
Specifications
| Back Material | Neoprene Padding |
| Color | Gray, Black |
| Hand | Specific |
| Size | 2X-Large |
| Glove Cuff Style | Hook and Loop |
| Palm Type | Tena-Grip™ Padding |
| Series | ProFlex™ 820 |