Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 760 Impact-Reducing Utility Work Gloves

Catalog No. 19820187 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-187 Small
19-820-188 Medium
19-820-189 Large
19-820-191 X-Large
19-820-192 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-187 Supplier Ergodyne™ Supplier No. 17662
This item is not returnable. View return policy

Thermoplastic rubber guards knuckles from impacts and scrapes, while padded palms are reinforced with Tena-Grip™ for additional protection.

  • TPR deflects knuckle busting impacts and scrapes
  • Tena-Grip™ reinforced palm, fingertips, and thumb saddle
  • Technical hex-stretch knit construction
  • Molded hook and loop closure
  • Gloves feature ID space to write user's names

Specifications

Color Black
Hand Specific
Material Thermoplastic Rubber
Product Type Work Gloves
Size Small
For Use With (Application) Heavy Duty Work, Construction, Mining, Oil and Gas, Mechanics, and Tool/Machine Operation
Glove Cuff Style Open
Series ProFlex™ 760
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.