Learn More
Description
- TPR deflects knuckle busting impacts and scrapes
- Tena-Grip™ reinforced palm, fingertips, and thumb saddle
- Technical hex-stretch knit construction
- Molded hook and loop closure
- Gloves feature ID space to write user's names
Specifications
Specifications
| Color | Black |
| Hand | Specific |
| Material | Thermoplastic Rubber |
| Product Type | Work Gloves |
| Size | X-Large |
| For Use With (Application) | Heavy Duty Work, Construction, Mining, Oil and Gas, Mechanics, and Tool/Machine Operation |
| Glove Cuff Style | Open |
| Series | ProFlex™ 760 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.