Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ ProFlex™ PCR System, 2 x Flat

Catalog No. p-7180372
Encompass_Preferred
Change view
Click to view available options
Warranty:
Limited Warranty
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Warranty
44-840-78 Limited Warranty
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-840-78 Supplier Applied Biosystems™ Supplier No. 4484078
Only null left
Add to Cart
Add to Cart
Request a Quote

The Veriti 96-well fast thermal cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex blocks, you have six independent temperature blocks that provide precise control over your PCR optimization.

The Veriti 96-well fast thermal cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex blocks, you have six independent temperature blocks that provide precise control over your PCR optimization. Additionally, the color touchscreen simplifies the setup and use of the Veriti 96-well fast thermal cycler. Optional setups for fast or standard PCR methods provide flexibility to shorten your PCR cycling times. The Veriti 96-well fast thermal cycler comes with a 0.1-mL block configuration that saves time and reagent consumption.

Features of the Veriti 96-well fast thermal cycler include:

  • Innovative VeriFlex blocks allow for precise PCR optimization or to run up to six independent assays
  • Six temperature zones for convenient PCR optimization
  • Standard- and fast-enabled cycling to address current and future PCR needs
  • Easy-to-use 6.5-inch VGA touchscreen
  • Convenient protocol transfer from one Veriti thermal cycler to another using USB
  • Reduced PCR cycling time when using faster ramp rates
  • Optional VeritiLink Remote Management Software allows remote management of more than 50 cyclers

VeriFlex blocks enhanced PCR functionality

Separate peltier blocks provide maximal versatility, flexibility, and offer two key benefits:

  1. Optimization—each block can be set with a specific temperature for precise control over PCR optimization
  2. More experiments—six peltier blocks can be used to run different annealing temperatures in the same run

Control at your fingertips

The powerful, yet simple to operate user interface on the Veriti system is driven by the 6.5-inch (16.51-cm) VGA color touchscreen. The large screen allows for easy viewing of temperature profiles, and large navigation buttons put programming at your fingertips. Setup and navigation of the Veriti Thermal Cycler does not require the use of a stylus or mouse.

Fast setup, fast results

The Veriti system has two options for method navigation. For quick setup, you may select one of the pre-programmed methods to run. If you prefer to program your own methods, simply touch the step you would like to setup and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.

Control your time, go fast when you want

With the Veriti Thermal Cycler you get two thermal cyclers in one: a standard thermal cycler with its better than gradient feature and a fast-enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti Thermal Cycler provides you with the flexibility to go fast when you want.

Applied Biosystems PCR plastics

Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment. To find the best match please visit our Plastics Selection Guide.

TRUSTED_SUSTAINABILITY

Specifications

Block Format Interchangeable
Thermal Accuracy ±0.25°C (35°C to 99.9°C)
High-throughput Compatibility High-throughput Compatible
Thermal Uniformity <0.5°C (20 sec. after reaching 95°C)
Reaction Speed Fast, Standard
Dimensions 22 x 13 x 10.6 in. (56.5 x 33 x 27 cm)
Capacity 8 OpenArray Plates, 24 x 20K Chips
Voltage 100/240 V
Display Type 8.4 in. Color TFT LCD Touchscreen
Product Type Thermal Cycler
Warranty Limited Warranty
Thermal Range 0°C to 100°C
For Use With (Application) PCR
Programs Auto re-start (after Power Outages), Program Overwrite Protection
User Interface USB, WiFi, Cloud, Ethernet
Max. Ramp Rate 3°C/sec. (Block), 1.6°C/sec. (Sample)
Quantity 1 Instrument
Memory USB and On-board
Weight 45 lb. (20.4 kg)
Block Configurations 2 x Flat Blocks
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.