Learn More
Description
- Breathable polyester mesh keeps hands cool without being bulky or stiff
- Low-profile, molded hook & loop closure provides a secure fit
- Gloves feature an ID tab for personalization to prevent misplaced gloves
- Pull-on tab for easy on/off
- Machine washable
Specifications
Specifications
| Coating Material | Silicone |
| Color | Black |
| Hand | Specific |
| Material | Polyester Mesh |
| Product Type | Work Gloves |
| Size | X-Large |
| For Use With (Application) | Warehousing/Distribution, Package Delivery |
| Glove Cuff Style | Open |
| Series | ProFlex™ 821 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.