Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 816 Thermal Fingerless Winter Work Gloves/Flip-Top Mittens

Catalog No. 19820159 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-159 Large
19-820-157 Small
19-820-158 Medium
19-820-160 X-Large
19-820-161 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-159 Supplier Ergodyne™ Supplier No. 17344
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mittens feature a soft microfleece lining for cold-weather protection and easily convert to half-finger gloves for detail work.

  • Mittens convert to half-finger gloves for detailed work
  • Flip-top mitten and thumb stow easily in mitten pocket when not in use
  • Soft microfleece lining provides comfort, flexibility, and protection in cold conditions
  • Rugged synthetic leather palm maintains flexibility in cold temps
  • Reinforced thumb saddle extends glove life
  • Contoured gathered elastic cuff with ID space
  • Gloves fit over jacket sleeves for convenient on/off wear
  • Reflective accents enhance wearer visibility and hand signaling
  • Soft thumb nose/brow wipe area

Specifications

Color Black
Hand Specific
Material Polyester
Size Large
EN388 Standard 3423 X
For Use With (Application) Construction, Cold Weather Detail Work, Cold Storage/Refrigeration, and Outdoor Recreation
Glove Cuff Style Elasticated
Liner Material Microfleece
Palm Type Synthetic Leather
Series ProFlex™ 816
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.