Learn More
Ergodyne™ ProFlex™ 812CR6 Utility + Cut-Resistant Gloves
Catalog No. 19820263 Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-820-263 Small
19-820-264 Medium
19-820-265 Large
19-820-266 X-Large
19-820-267 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-820-263 Supplier Ergodyne™ Supplier No. 17922
Add to Cart
Add to Cart
This item is not returnable. View return policy
Built for protection from extreme cut hazards without sacrificing dexterity. Synthetic leather palm and reinforced thumb saddle provide enhanced grip and feel when handling materials.
  • Next-gen Armortex™ delivers ANSI/ASTM Level A6 cut protection
  • Contrasting fingertips for hand signaling
  • Synthetic leather palm with reinforced thumb saddle for grip and feel
  • Breathable poly mesh keeps hands cool and is not bulky or stiff
  • Low-profile, molded hook and loop closure
  • Machine washable
Specifications
Coating Material Leather
Color Black, Red
Cut Resistance Rating ANSI A6
Material Poly Mesh
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:2543 E
For Use With (Application) Assembly/Fabrication, Sheet Metal, Heavy Construction, Oil and Gas, Sharp Edge Material Handling, Electrical, Pulp and Paper, Service
Model 17922
Quantity 1 Pair/Pk., 6 Pk./Cs.
Series ProFlex™ 812CR6
Thumb Style Reinforced