Learn More
Description
- Next-gen Armortex™ delivers ANSI/ASTM Level A6 cut protection
- Contrasting fingertips for hand signaling
- Synthetic leather palm with reinforced thumb saddle for grip and feel
- Breathable poly mesh keeps hands cool and is not bulky or stiff
- Low-profile, molded hook and loop closure
- Machine washable
Specifications
Specifications
| Coating Material | Leather |
| Color | Black, Red |
| Cut Resistance Rating | ANSI A6 |
| Material | Poly Mesh |
| Product Type | Cut-resistant Gloves |
| Size | Large |
| EN388 Standard | EN 388:2543 E |
| For Use With (Application) | Assembly/Fabrication, Sheet Metal, Heavy Construction, Oil and Gas, Sharp Edge Material Handling, Electrical, Pulp and Paper, Service |
| Model | 17924 |
| Quantity | 1 Pair/Pk., 6 Pk./Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.