Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Progerin Monoclonal Antibody (13A4D4)
GREENER_CHOICE

Catalog No. PIMA518073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518073 Supplier Invitrogen™ Supplier No. MA518073
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody specifically recognizes Progerin (Lamin A delta50) and does not cross-react with endogenous prelamin A. Western blot results show a band at approximately 70 kDa.

Progerin is a 614 amino acid protein involved in Hutchinson-Gilford progeria syndrome. Progerin is most often generated by a point mutation (C1824T) in the LMNA gene that codes for lamin-A/C. This mutation activates a cryptic splice site and gives rise to a form of lamin A with a 50-amino acid internal deletion within the C-terminal domain of the protein. Approximately 80% of Hutchinson-Gilford progeria syndrome cases carry a single copy of the most common mutation, a silent point mutation, G608G (GGC > GGT), within exon 11 of the LMNA gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Progerin
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 13A4D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LMNA
Gene Accession No. P02545
Gene Alias 70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A/C; lamin A/C-like 1; Lamin AC; Lamin-A/C; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; Mutant lamin A/C; Prelamin-A/C; PRO1; progerin mutant; Renal carcinoma antigen NY-REN-32
Gene Symbols LMNA
Host Species Mouse
Immunogen Synthetic peptide conjugated to KLH derived from within residues 604-611 (GAQSPQNC) at the C terminus of Human Progerin.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.