Learn More
U.S. Pharmacopeia Progesterone, 57-83-0, MFCD00003658, 200mg
Supplier:  U.S. Pharmacopeia 1568007
Encompass
Molecular formula C21-H30-O2, Molecular weight 314.47 g/mol, Melting point/freezing point 260.6 - 267.8 °F (127 - 131 °C), Synonyms: Corpus luteum hormone
Catalog No. NC9991433
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.