Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Progesterone Competitive ELISA Kit

Catalog No. EIAP4C21
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAP4C21 96 Tests
EIAP4C21X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EIAP4C21 Supplier Invitrogen™ Supplier No. EIAP4C21
Add to Cart
Add to Cart

ELISA

The Progesterone ELISA quantitates progesterone in urine,dried fecal extracts or cell culture medium.

Principle of the method

The Progesterone Competitive ELISA research-use-only kit is designed to quantitatively measure Progesterone present in extracted dried fecal samples,urine,and tissue culture media samples. A progesterone standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture mouse antibodies. A progesterone-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a monoclonal antibody to progesterone to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound progesterone-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

Progesterone is the major progestational steroid, secreted primarily by the corpus luteum and the placenta. Progesterone binds to the progesterone receptor, resulting in dissociation of heat shock proteins, receptor phosphorylation, and transcription activation through direct or indirect interaction with transcription factors. Progesterone acts on the uterus, the mammary glands and the brain. It is required in embryo implantation, pregnancy maintenance, and the development of mammary tissue for milk production. Progesterone, converted from pregnenolone, also serves as an intermediate in the biosynthesis of gonadal steroid hormones and adrenal corticosteroids. Therapeutic progesterone is a synthetic form of the endogenous hormone progesterone. This agent exerts inhibitory effects on estrogens by decreasing the number of estrogen receptors and increasing its metabolism to inactive metabolites. [National Center for Biotechnology Information. PubChem Compound Database; CID=5994]
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 50 to 3200 pg/mL
Assay Sensitivity 47.9 pg/mL
Description Progesterone Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Steroid hormone
Sample Type Fecal Extract, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 5.7%
Intraassay CV 3.9
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Progesterone
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.