Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Progesterone, Endogenous Progesterone Receptor Agonist, Solid, 100 mg

Catalog No. 502706758
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6758 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6758 Supplier Abcam Supplier No. AB141252100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-21 steroid hormone serving as an endogenous progesterone receptor agonist.

  • Molecular Weight: 314.5 Da
  • Purity: >99%
  • Description: Endogenous progesterone receptor agonist (EC50 = 0.5 nM) C-21 steroid hormone; elicits a variety of biological effects
  • Form: Solid
  • Composition: 100% Progesterone

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Progesterone Receptor
Substrate Properties Chemical Substrate
Buffer 100% Progesterone
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Quantity 100 mg
For Use With (Application) Progesterone receptor agonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.