Learn More
Description
MA1-12626 detects Progesterone Receptor in human, equine and porcine samples. MA1-12626 has been successfully used in immunofluorescence, immunoprecipitation, western blot and immunohistochemistry (paraffin, frozen) procedures. The MA1-12626 immunogen is human endometrial carcinoma from in athymic mice.
Specifications
Specifications
| Antigen | Progesterone Receptor |
| Applications | ChIP Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Cocktail |
| Clone | hPRa2, hPRa3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | PGR |
| Gene Accession No. | P06401 |
| Gene Alias | 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.